Learn More
Enzo Life Sciences HSC 70 HSP 73 (bovine), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP751F
Encompass_Preferred
Alternative name: Heat shock cognate 71 kDa protein, Heat shock 70 kDa protein 8, HSPA8. MW: ~73kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In 30mM TRIS-HCl, pH 7.5, containing 1.0mM EDTA, 2.0mM DTT, and 300mM sodium chloride. Source: Produced in E. coli. UniProt: P19120. Application: WB, Activity assay. Long Term Storage: -80°C
Catalog No. 50-667-756
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.