Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103192 Supplier Invitrogen™ Supplier No. PA5103192
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DHRS8.

Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSD17B11
Gene Accession No. Q8NBQ5, Q9EQ06
Gene Alias 17-beta-HSD 11; 17-beta-HSD XI; 17betaHSD11; 17-BETA-HSD11; 17beta-HSD11; 17betaHSDXI; 17-BETA-HSDXI; 17-beta-hydroxysteroid dehydrogenase 11; 17-beta-hydroxysteroid dehydrogenase type XI; 17-beta-hydroxysteroid dehydrogenase XI; 17bHSD11; CTCL tumor antigen HD-CL-03; CTCL-associated antigen HD-CL-03; Cutaneous T-cell lymphoma-associated antigen HD-CL-03; dehydrogenase/reductase (SDR family) member 8; dehydrogenase/reductase SDR family member 8; Dhrs8; estradiol 17-beta-dehydrogenase 11; estradiol 17-beta-dehydrogenase 11-like protein; Hsd17b11; hydroxysteroid (17-beta) dehydrogenase 11; hydroxysteroid 17-beta dehydrogenase 11; MGC137132 protein; PAN1B; PSEC0029; Retinal short-chain dehydrogenase/reductase 2; retinal short-chain dehydrogenase/reductase SDR2; retSDR2; SDR16C2; SDR2; short chain dehydrogenase/reductase family 16C member 2; short chain dehydrogenase/reductase family 16C, member 2; UNQ207/PRO233
Gene Symbols HSD17B11
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSD17B11(Accession Q8NBQ5), corresponding to amino acid residues K212-S262.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114664, 51170
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.