Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501731498 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1498 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1498 Supplier Proteintech Group Inc Supplier No. 173011AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.

Specifications

Antigen HSD17B11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HSD17B11
Gene Accession No. Q6AYS8, Q8NBQ5, Q9EQ06
Gene Alias 17 beta HSD 11, 17 beta HSD XI, 17 BETA HSD11, 17 BETA HSDXI, 17betaHSD11, 17betaHSDXI, 17bHSD11, DHRS8, HSD17B11, PAN1B, RETSDR2, SDR16C2, UNQ207/PRO233
Gene Symbols Hsd17b11
Host Species Rabbit
Immunogen HSD17B11 Fusion Protein Ag11182
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114664, 289456, 51170
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.