Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109834 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109834 Supplier Invitrogen™ Supplier No. PA5109834
  • $588.00
    $485.65 / Each of 1

    Save $102.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: RRKSVAGEIV LITGAGHGIG RQTTYEFAKR QSILVLWDIN KRGVEETAAE CRKLGVTAHA YVVDCSNREE IYRSLNQVKK EVGDVTIVVN NAGTVYPADL LSTKDEEITK TFEVNILGHF WITKALLPSM MERNHGHIVT VASVCGHEGI PYLIPYCSSK FAAVGFHRGL TSELQALGKT GI.

HSD17B13 (Hydroxysteroid 17 Beta Dehydrogenase 13) is a Protein Coding gene. Diseases associated with HSD17B13 include5 Alpha Reductase DeficiencyandMucopolysaccharidosis Iii. Among its related pathways areMetabolismandLipoprotein metabolism. Gene Ontology (GO) annotations related to this gene includeoxidoreductase activity. An important paralog of this gene isHSD17B11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B13
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene HSD17B13
Gene Accession No. Q5M875, Q7Z5P4, Q8VCR2
Gene Alias 17-beta hydroxysteroid dehydrogenase; 17-beta hydroxysteroid dehydrogenase 13; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; AI047820; Alcohol dehydrogenase PAN1B-like; HMFN0376; Hsd17b13; hydroxysteroid (17-beta) dehydrogenase 13; hydroxysteroid 17-beta dehydrogenase 13; NIIL497; Pan1b; PAN1B-like; SCDR9; SDR16C3; short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; short-chain dehydrogenase/reductase 9; UNQ497/PRO1014
Gene Symbols HSD17B13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-211 of human HSD17B13 (NP_835236.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243168, 305150, 345275
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.