Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B3 Monoclonal Antibody (OTI7F7), TrueMAB™, OriGene
SDP

Catalog No. p-200017657
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4087 Supplier OriGene Supplier No. TA811500
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predomitly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia.

Specifications

Antigen HSD17B3
Applications Western Blot
Classification Monoclonal
Clone OTI7F7
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HSD17B3
Gene Accession No. P37058
Gene Alias EDH17B3, SDR12C2
Gene Symbols HSD17B3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 49-268 of human HSD17B3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3293
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.