Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSD17B7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143945
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143945 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143945 Supplier Invitrogen™ Supplier No. PA5143945
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human HL-60 whole cell, human 293T whole cell, human Jurkat whole cell, human RT4 whole cell, rat liver tissue, rat lung tissue, rat RH35 whole cell. IHC: human liver cancer tissue, human prostate cancer tissue, human colorectal adenocarcinoma tissue. ICC/IF: HepG2 cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Responsible for the reduction of the keto group on the C-3 of sterols.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSD17B7
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene HSD17B7
Gene Accession No. P56937, Q62904
Gene Alias 17 beta-hydroxysteroid dehydrogenase type VII; 17beta hydroxysteroid dehydrogenase; 17-beta-HSD 7; 17-beta-hydroxysteroid dehydrogenase 7; 3-keto-steroid reductase; AI266814; bA291L22.1; ERG27; Estradiol 17-beta-dehydrogenase 7; Hsd17b_2; HSD17B7; HSD17B7P2; hydroxysteroid (17-beta) dehydrogenase 7; hydroxysteroid 17-beta dehydrogenase 7; hydroxysteroid 17-beta dehydrogenase 7 pseudogene 2; hydroxysteroid dehydrogenase 17 beta, type 7; PRAP; PRL receptor-associated protein; SDR37C1; Short chain dehydrogenase/reductase family 37C member 1; short chain dehydrogenase/reductase family 37C, member 1; UNQ2563/PRO6243
Gene Symbols HSD17B7
Host Species Rabbit
Immunogen E.coli-derived human HSD17B7 recombinant protein (Position: L16-L336).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29540, 51478
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.