Learn More
AGI NORTH AMERICA INC HS Martin 50L spherical distillation system installed in a corrosion resistant stationary framework, electrically heated, equipped with a 3” dia x 36” column, continuous liquid separator, 4.3ft² (.4m²) condenser and 22L receiving flask
SDP
Supplier:  AGI NORTH AMERICA INC 5003600050F
Encompass_Preferred
System pressure rating: up to 7PSI (.5Bar) to full vacuum-System is rated for to handle continuous temperatures up to +200 deg C-Framework constructed with polished SS pipe mounted on extra heavy feet for extra stability-All glassware is constructed of 33 expansion borosilicate glass-50L flask has 5 necks: 100mm center neck, one 80mm side neck for column, one 80mm side neck for addition/cleaning. Two 40mm side necks for accessories: one thermowell and one dip tube included-All wetted surfaces are constructed of either borosilicate glass or PTFE-Supplied o-rings are FKM, other compounds are available upon request-Flask is heated by an aluminum-housed bottom high-temperature 230V mantle that has three circuits rated at 2,000 watts each-Instrumentation and controls can be provided upon request - please contact one of our technical service advisors to discuss optional equipment available for your specific application
Catalog No. NC2417600
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.