Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP105 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536059 Supplier Invitrogen™ Supplier No. PA536059
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 105 kDa. Purity is >95% by SDS-PAGE.

HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP105
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene HSPH1
Gene Accession No. Q61699, Q66HA8, Q92598
Gene Alias 105kDa; 42 degrees C-HSP; AI790491; antigen NY-CO-25; heat shock 105/110 protein 1; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa; heat shock 105kDa/110kDa protein 1; heat shock 110 kDa protein; Heat shock protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein family H (Hsp110) member 1; heat shock protein, 105 kDa; heat shock-related 100 kDa protein E7I; heat-shock protein 110 kDa; HSP; Hsp105; HSP105 42 C-HSP; HSP105A; HSP105B; Hsp110; hsp110/105; HSP-E7I; Hsph1; I79_002075; Kiaa0201; KIAA0202; NY CO 25; NY-CO-25; SEP2; SEPT8; septin 8; SEPTIN8; septin-8
Gene Symbols HSPH1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 780-830 of human HSP105.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10808, 15505, 288444
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.