Learn More
Invitrogen™ HSP40 Monoclonal Antibody (OTI1E9)

Description
HSP40 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | HSP40 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1E9 |
| Concentration | 1.00 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DNAJB1 |
| Gene Accession No. | P25685 |
| Gene Alias | 0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.