Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP60 Monoclonal Antibody (4B9/89), Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIMA3012AY Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3012AY 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3012AY Supplier Invitrogen™ Supplier No. MA3012A647
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The MA3012A647 antigen is human placental HSP60. Epitope mapping studies using human HSP60 deletion mutants suggest that this antibody binds either between amino acids 335-366 or 484-547.

This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP60
Applications Immunocytochemistry
Classification Monoclonal
Clone 4B9/89
Concentration 1 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene HSPD1
Gene Accession No. P10809, P63038, P63039
Gene Alias 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409)
Gene Symbols HSPD1
Host Species Mouse
Immunogen Human placental HSP60.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15510, 3329, 63868
Target Species Human, Mouse, Monkey, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.