Learn More
Selleck Chemical LLC HSP990 (NVP-HSP990) 5mg 934343-74-5
SDP
Supplier:  Selleck Chemical LLC S70975MG
Encompass_Preferred
HSP990 (NVP-HSP990) is a novel, potent and selective HSP90 inhibitor for HSP90?/? with IC50 of 0.6 nM/0.8 nM. NVP-HSP990 induces cell cycle arrest and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.