Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPB7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569143 Supplier Invitrogen™ Supplier No. PA569143
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 90%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 85%; Rat: 92%.

HSPB7 encodes a small heat shock family B member that can heterodimerize with similar heat shock proteins. Defects in this gene are associated with advanced heart failure. In addition, the encoded protein may be a tumor suppressor in the p53 pathway, with defects in this gene being associated with renal cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPB7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HSPB7
Gene Accession No. P35385
Gene Alias 27kDa; Cardiovascular heat shock protein; cvHsp; heat shock 27kD protein family member 7; heat shock 27kD protein family, member 7 (cardiovascular); heat shock 27kDa protein family member 7 (cardiovascular); heat shock 27kDa protein family, member 7 (cardiovascular); Heat shock protein; Heat shock protein 25 kDa 2; heat shock protein 25 kDa 2 (cardiovascular); heat shock protein B7; heat shock protein beta-7; heat shock protein family B (small) member 7; heat shock protein family, alpha-crystallin-related, b7; heat shock protein family, member 7 (cardiovascular); HSP; Hsp25-2; HspB7; Protein p19/6.8; zgc:103426
Gene Symbols HSPB7
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse HSPB7 (aa 51-100).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29818
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.