Learn More
HSPBP1 Mouse anti-Human, Clone: OTI3E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503554

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
HspBP1 is an Hsp70-interacting protein that was first isolated from a human heart cDNA library using the yeast two-hybrid system (1,2). The ATPase domain of Hsp70 binds HspBP1 and upon binding, inhibits Hsp70 chaperone activity. More recently HspBP1 was found to promote nucleotide dissociation from Hsc70 (3) and is capable of altering the confirmation of the Hsp70 ATPase domain (4). HspBP1 is capable of regulating any Hsp70 activity that requires ATPase.Specifications
HSPBP1 | |
Monoclonal | |
Unconjugated | |
HSPBP1 | |
FES1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23640 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI3E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NZL4 | |
HSPBP1 | |
Full length human recombit protein of human HSPBP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.