Learn More
Creative Biomart Recombinant HSV-2 gD Antigen, His-tagged
Supplier:  Creative Biomart HSV239
Encompass_Preferred
The E. coli derived recombinant protein contains the HSV-2 gD (525-578) immunodominant region.HSV2 gD is fused to a six histidine his tag at c-terminus and purified by proprietary chromatographic tech
Catalog No. 50-208-4617
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.