Learn More
Diagnostic Biosystems HSV I Herpes Simplex Virus Type I (HSV1) - IVD
Supplier: Diagnostic Biosystems RP01805
This antibody reacts with HSV type I specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.