Learn More
Polyscience 15L Heated Circulator, Advanced Digital Controller (Ambient +10° to 200°C); 120V, 60Hz
SDP
Supplier:  Polyscience AD15H200A11B
Encompass_Preferred
PolyScience™ 15 Liter Heated CirculatorAdvanced Digital ControllerAmbient +10 to 200°C Working Temperature RangeTemperature Stability: ±0.01°1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate: 5.3 gpm (20.1 LPM) Maximum Suction Flow Rate: 3.9 gpm (14.7 LPM)120V, 60HzFeatures:• Intuitive 3.75” (9.5 cm) display with touch-pad control• 4 languages: French, German, Spanish, English• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485 and external temperature probe• On-screen prompts• Automatic performance optimization and specific heat tuning• Single-point calibration capability
Catalog No. 50-113-0623
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.