Learn More
Polyscience 7L Heated Circulator, MX Controller (Ambient +10 to 135°C); 120V, 60Hz
SDP
Supplier:  Polyscience MX07H135A11B
Encompass_Preferred
PolyScience™ 7 Liter Heated CirculatorMX ControllerAmbient +10 to 135°C Working Temperature RangeTemperature Stability: ±0.07°C1100W HeaterMaximum Pressure: 2.3 psi (0.16 bar) Maximum Pressure Flow Rate: 3.6 gpm (13.5 LPM) 120V, 60HzFeatures:• Large, universal icon and English display• On-screen prompts• Single-point calibration capability
Catalog No. 50-113-0634
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.