Learn More
Foxx Life Sciences CPC AseptiQuik S HT Connector, 1/4" Hose Barb Genderless, Polycarbonate (PC), Class VI, AQS17004HT, 100/PK
Supplier:  Foxx Life Sciences AQS17004HT100
Encompass_Preferred
Aseptiquik S Connectors Small Format 1/8", 1/4" and 3/8" hose barb and the 1/4" & 3/4" sanitary termination Connectors provide quick and easy sterile connections for small-flow applications, even in non-sterile environments. The “FLIP-CLICK-PULL” design of AseptiQuik S enables users to quickly transfer small volumes of media easily with less risk of operator error. The connector’s genderless and robust design provides reliable performance without the need for clamps, fixtures, or tube welders. Biopharmaceutical manufacturers can now make 1/8", 1/4" and 3/8" sterile connections with the quality and market availability they expect from the leader in single-use connection technology.
Catalog No. 50-206-0561
Add to Cart
Request a Quote