Learn More
Description
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
- Do not use past expiration date.
- Do not mix or substitute reagents with those from other lots or sources.
- Variations in sample collection, processing, and storage may cause sample value differences.
- When mixing or reconstituting protein solutions, always avoid foaming.
- To avoid cross-contamination, change pipette tips between additions of each standard level, between sample additions, and between reagent additions. Also, use separate reservoirs for each reagent.
- To ensure accurate results, proper adhesion of plate sealers during incubation steps is necessary.
- The acute and chronic effects of overexposure to reagents in this kit are unknown. Wear protective gloves, clothing, eye, and face protection. Wash hands thoroughly after handling. Refer to the SDS on our website prior to use.
- High throughput ELISA to quantify γ-H2AX in human cultured cells, peripheral blood mononuclear cells, and tissue biopsies.
- In this assay, immobilized γ-H2AX antibody in the wells of a 96-well plates is used to capture γ-H2AX from sample lysate.
Specifications
Specifications
| Target Species | Multi-species |
| Gene ID (Entrez) | 3014 |
| Quantity | 96 Tests |
| Storage Requirements | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Synonym | H2A.X, H2A/X, H2AFX |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
