Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HTH 01-015
GREENER_CHOICE

Catalog No. p-7110850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-221-0 10 mg
56-225-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-221-0 Supplier Tocris Bioscience™ Supplier No. 5622/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NUAK1 inhibitor

HTH 01-015 is a potent and selective NUAK1 inhibitor (IC50 = 100 nM). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells in vitro, to a similar extent as NUAK1 knockdown models.

Specifications

Chemical Name or Material 5,13-Dihydro-4,5,13-trimethyl-2-[[1-(4-piperidinyl)-1H-pyrazol-4-yl]amino]-6H-naphtho[2,3-e]pyrimido[5,4-b][1,4]diazepin-6-one
CAS 1613724-42-7
Quantity 10 mg
Target Other Kinase Inhibitors
Molecular Formula C26H28N8O
Formula Weight 468.55
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.