Learn More
Chemglass Life Sciences 3000mL Glas-Col Heating Mantle, Series O, Hemispherical, 500 Watts
SDP
Catalog No. CG1000010
Encompass_Preferred
3000mL Glas-Col Heating Mantle, Series O, Hemispherical, 500 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.
Catalog No. CG1000010 Supplier Chemglass Life Sciences Supplier No. CG1000010
3000mL Glas-Col Heating Mantle, Series O, Hemispherical, 500 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.