Learn More
Invitrogen™ HTR1B Polyclonal Antibody

Description
Suggested positive control: human, mouse, or rat brain.
Specifications
Specifications
| Antigen | HTR1B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | HTR1B |
| Gene Accession No. | P28222, P28334, P28564 |
| Gene Alias | 5-HT>1B> receptor; 5ht1b; 5-HT1B; 5-HT-1B; 5HT1B Receptor; 5-HT1B receptor; 5-HT1B serotonin receptor; 5-HT1DB; 5-HT-1D-beta; 5-hydroxytryptamine (serotonin ) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; HTR1B; HTR1D2; HTR1DB; S12; serotonin (5-hydroxytryptamine (5HT)) receptor type 1B; serotonin 1D beta receptor; Serotonin receptor 1B |
| Gene Symbols | HTR1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.