Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112579 Supplier Thermo Scientific Supplier No. PA5112579
Only null left
Add to Cart
Add to Cart

HTR3B Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR3B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HTR3B
Gene Accession No. O95264, Q9JHJ5
Gene Alias 5-HT3 receptor subunit B; 5-HT3B; 5-HT-3B; 5-HT3-B; 5-HT3B receptor; 5-hydroxytryptamine (serotonin) receptor 3B; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; HTR3B; Serotonin receptor 3B; serotonin-gated ion channel subunit
Gene Symbols HTR3B
Host Species Rabbit
Immunogen Recombinant Human 5-hydroxytryptamine receptor 3B protein (22-238AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57014, 9177
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.