Learn More
HTRA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The 50 kDa HTRA1 was found at varying levels in the majority of samples tested, whereas the 42 kDa form of HTRA1 was identified in degenerated intervertebral disc protein samples. The antibody is specific to HTRA1.
Specifications
Specifications
| Antigen | HTRA1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HTRA1 |
| Gene Accession No. | Q92743, Q9QZK5, Q9R118 |
| Gene Alias | ARMD7, HtrA, HtrA serine peptidase 1, HTRA1, L56, ORF480, PRSS11, Serine protease 11, Serine protease HTRA1 |
| Gene Symbols | Htra1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.