Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTT Mouse anti-Human,Mouse, Unconjugated, Clone: MW8, MilliporeSigma™

Catalog No. p-7181790 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25292MI 25 μg
MABN25291MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25292MI Supplier MilliporeSigma Supplier No. MABN252925UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HTT Monoclonal antibody specifically detects HTT in Human,Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTT
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MW8
Conjugate Unconjugated
Gene Accession No. NP_002102.4
Gene Alias Huntingtin;Huntington disease protein:HD protein
Gene Symbols HTT, HD, IT15
Host Species Mouse
Immunogen HD exon-1 67Q aggregate.
Molecular Weight of Antigen 347.6 kDa calclulated
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone MW8 is a mouse monoclonal antibody that detects Huntingtin protein. It targets an epitope within nine amino acids in the exon-1 region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.