Learn More
Bon Opus Biosciences Recombinant Human Brain-Derived Neurotrophic Factor/BDNF 1mg
SDP
Supplier:  Bon Opus Biosciences C0761MG
Encompass_Preferred
Recombinant Human Brain-Derived Neurotrophic Factor is produced by our E.coli expression system and the target gene encoding His129-Arg247 is expressed. Accession# P23560. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 250mM NaCl, pH 7.2. Bon Opus Cat# C076
Catalog No. 50-217-2721
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.