Learn More
Bon Opus Biosciences Recombinant Human IL-2RB/CD122 (C-hFc) -500ug
SDP
Supplier:  Bon Opus Biosciences BP150500UG
Encompass_Preferred
Recombinant Human Interleukin-2 Receptor subunit beta is produced by our Mammalian expression system and the target gene encoding Ala27-Asp239 is expressed with a Fc tag at the C-terminus. Expression system: Human 293 Cells. Accession# P14784. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP150
Catalog No. 50-208-0238
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.