Learn More
Bon Opus Biosciences Recombinant Human IL-3R alpha/CD123(C-hFc) -500ug
SDP
Supplier:  Bon Opus Biosciences BP123500UG
Encompass_Preferred
Recombinant Human Interleukin-3 Receptor Subunit Alpha is produced by our Mammalian expression systemand the target gene encoding Lys20-Arg305 is expressed with a Fc tag at the C-terminus. Expression system: Human 293 Cells. Accession# P26951. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP123
Catalog No. 50-208-0197
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.