Learn More
Bon Opus Biosciences Biotinylated Recombinant Human IL-2RA/CD25 (C-Avi/His) -250ug
SDP
Supplier:  Bon Opus Biosciences BP136250UG
Encompass_Preferred
Biotinylated recombinant Human Interleukin-2 Receptor subunit alpha is produced by our Mammalian expression system and the target gene encoding Glu22-Gln240 is expressed with a Avi and 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P01589. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP136
Catalog No. 50-208-0221
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.