Learn More
Bon Opus Biosciences Recombinant Human CD73 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW8450UG
Encompass_Preferred
Recombinant Human 5'-Nucleotidase is produced by our Mammalian expression system and the target gene encoding Trp27-Lys547 is expressed. Expression system: Human Cells. Accession# P21589. Formulation: Supplied as a 0.2 um filtered solution of 20mMTrisHCl,120mMNaCl,4mMCaCl2,20%Glycerol,pH7.5. Bon Opus Cat# CW84
Catalog No. 50-208-0464
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.