Learn More
Bon Opus Biosciences Recombinant Human CELA3A(C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP17250UG
Encompass_Preferred
Recombinant Human Chymotrypsin-Like Elastase Family Member 3A is produced by our Mammalian expression system and the target gene encoding Ser16-His270 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P09093. Formulation: Supplied as a 0.2 um filtered solution of 20mM Tris-HCl, 150mM NaCl, 10%Glycerol, pH7.5. Bon Opus Cat# BP172
Catalog No. 50-208-0273
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.