Learn More
Raybiotech Inc Human Cell Cycle Array L1, Glass Slide, 8 Sample Kit
Supplier:  Raybiotech Inc AAHBLGCYC8
Encompass_Preferred
Label-based antibody array kit using glass slide supports to detect 89 human proteins involved in cell cycle pathways. Suitable for serum, plasma, lysates, and cell culture supernatants.
Catalog No. NC2825061