Learn More
Bon Opus Biosciences Recombinant Human COQ7 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP03250UG
Encompass_Preferred
Recombinant Human 5-demethoxyubiquinone hydroxylase, mitochondrial is produced by our Mammalian expression system and the target gene encoding Ser37-Leu217 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q99807. Formulation: Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP032
Catalog No. 50-208-0118
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.