Learn More
Bon Opus Biosciences Recombinant Human ER alpha (N-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP15750UG
Encompass_Preferred
Recombinant Human Estrogen receptor alpha is produced by our E.coli expression system and the target gene encoding Met1-Gln116 is expressed with a 6His tag at the N-terminus. Expression system: E. coli. Accession# P03372. Formulation: Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# BP157
Catalog No. 50-208-0247
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.