Learn More
Bon Opus Biosciences Recombinant Human Ferritin heavy chain/FTH -50ug
SDP
Supplier:  Bon Opus Biosciences BP15950UG
Encompass_Preferred
Recombinant Human Ferritin heavy chain is produced by our E.coli expression system and the target gene encoding Met1-Ser183 is expressed. Expression system: E. coli. Accession# P02794. Formulation: Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP159
Catalog No. 50-208-0251
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.