Learn More
Bon Opus Biosciences Recombinant Human G-CSFR (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW3550UG
Encompass_Preferred
Recombinant Human Granulocyte Colony-stimulating Factor Receptor is produced by our Mammalian expression system and the target gene encoding Glu25-His627 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q99062. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW35
Catalog No. 50-208-0420
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.