Learn More
MyBioSource HARS Human ELISA Kits, Histidyl-tRNA Synthetase
SDP
Supplier:  MyBioSource MBS90290196STRIPWELLS
Encompass_Preferred
The MyBioSource Human Histidyl-tRNA Synthetase ELISA Kit quantitatively measures this enzyme in various biological fluids using a sandwich immunoassay technique. The kit, with a 0.156-10 ng/mL range, includes pre-coated 96-well plates, detection antibodies, HRP conjugate, TMB substrate, and necessary protocols.
Catalog No. NC2607137
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.