Learn More
Bon Opus Biosciences Recombinant Human Interleukin-2/IL2 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP059500UG
Encompass_Preferred
Recombinant Human Interleukin-2 is produced by Human 293 Cells. The target gene encoding A21-T153 is expressed with a 6His tag at the C terminus. Expression system: Human 293 Cells. Accession# P60568. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP059
Catalog No. 50-208-0134
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.