Learn More
Bon Opus Biosciences Recombinant Human Interferon regulatory factor 5/IRF5 (C-His) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP25650UG
Encompass_Preferred
Recombinant Human Interferon Regulatory Factor 5 is produced by our E.coli expression system and the target gene encoding Met1-Gln498 is expressed with a 6His tag at the C-terminus. Accession#Q13568. Formulation: Supplied as a 0.2 um filtered solution of 20mM Tris-HCl, 150mM NaCl, 30% Glycerol, 1mM DTT, pH8.0
Catalog No. NC2733738
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.