Learn More
Bon Opus Biosciences Recombinant Human Kallikrein 5/KLK5 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP127500UG
Encompass_Preferred
Recombinant Human Kallikrein 5 is produced by our Mammalian expression system and the target gene encoding Val23-Ser293 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q9Y337. Formulation: Supplied as a 0.2 um filtered solution of 20mM MES, 150mM NaCl, 10% Glycerol, pH 5.5. Bon Opus Cat# BP127
Catalog No. 50-208-0203
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.