Learn More
Bon Opus Biosciences Recombinant Human Leptin Receptor/LEPR/CD295 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP148500UG
Encompass_Preferred
Recombinant Human Leptin Receptor is produced by our Mammalian expression system and the target gene encoding Phe22-Asp839 is expressed with a 10His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P48357. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP148
Catalog No. 50-208-0234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.