Learn More
Bon Opus Biosciences Recombinant Human OX40L (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW2550UG
Encompass_Preferred
Recombinant Human OX40 Ligand is produced by our Mammalian expression system and the target geneencoding Gln51-Leu183 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# P23510. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CW25
Catalog No. 50-208-0414
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.