Learn More
Bon Opus Biosciences Biotinylated Recombinant Human Polymeric Immunoglobulin Receptor/PIgR (C-His) -250ug
SDP
Supplier:  Bon Opus Biosciences BP075250UG
Encompass_Preferred
Biotinylated Recombinant Human Polymeric Immunoglobulin Receptor is produced by Human 293 Cells. The target gene encoding K19-R639 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P01834. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.6. Bon Opus Cat# BP075
Catalog No. 50-208-0150
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.