Learn More
Bon Opus Biosciences Recombinant Human Serpin B3/SCCA-1 (C-Avi/His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP089500UG
Encompass_Preferred
Recombinant Human Serpin B3 is produced by E.coli. The target gene encoding M1-P390 is expressed with a Avi/;6His tag at the C terminus. Expression system: E. coli. Accession# P29508. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM Na2HPO4, 2umM NaH2PO4, pH7.4. Bon Opus Cat# BP089
Catalog No. 50-208-0164
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.