Learn More
Bon Opus Biosciences Recombinant Human Sulfamidase/SGSH (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP124500UG
Encompass_Preferred
Recombinant Human N-Sulphoglucosamine Sulphohydrolase is produced by our Mammalian expression system and the target gene encoding Arg21-Leu502 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P51688. Formulation: Supplied as a 0.2 um filtered solution of 20mM TrisHCl,150mM NaCl,1mM GaCl2,10%Glycerol,pH7.5. Bon Opus Cat# BP124
Catalog No. 50-208-0198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.