Learn More
Bon Opus Biosciences Recombinant Human Siglec-2 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CC0350UG
Encompass_Preferred
Recombinant Human B-cell Receptor CD22 is produced by our Mammalian expression system and the target gene encoding Asp20-Arg687 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# P20273. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,1mM EDTA,pH7.4. Bon Opus Cat# CC03
Catalog No. 50-208-0379
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.