Learn More
Bon Opus Biosciences Biotinylated Recombinant Human Thioredoxin/TXN (C-Avi/His) -250ug
SDP
Supplier:  Bon Opus Biosciences BP092250UG
Encompass_Preferred
Biotinylated Recombinant Human Thioredoxin is produced by E.coli. The target gene encoding M1-V105 is expressed with a Avi/;6His tag at the C terminus. Expression system: E. coli. Accession# P10600. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM Na2HPO4, 2umM NaH2PO4, pH7.4. Bon Opus Cat# BP092
Catalog No. 50-208-0167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.