Learn More
Invitrogen™ HuC/HuD Monoclonal Antibody (16A11), Biotin
Description
This antibody recognizes the Elav family members HuC, HuD and Hel-N1 neuronal proteins. It does not recognize HuR, another Elav family member that is present in all proliferating cells. The antibody has been shown to specifically label neuronal cells in zebrafish, chick, canaries, and humans, and is likely to label neuronal cells in most vertebrate species. Labeling is visible early in development, at about the time that the neurons leave the mitotic cycle. Storage and reconstitution: Upon receipt, lyophilized antibodies should be stored desiccated at -20°C or lower. When properly stored, these products are stable for at least one year. To prepare stock solution, reconstitute the antibody in 0.5-1 mL PBS, pH 7.4, containing 1% BSA. Store the solution for up to two weeks at 4°C with the addition of 2mM sodium azide. For longer storage, divide solutions into single-use aliquots and freeze at -20°C, avoidig freeze/thaw cycles. For best results in immunohistochemistry, antigen retrieval is required. Use 50mM Tris, pH 8.0, for 30 minutes. The biotin-XX labeled antibody can be detected by using fluorescently labeled avidin, streptavidin or NeutrAvidin conjugates.
Specifications
Specifications
| Antigen | HuC/HuD |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 16A11 |
| Conjugate | Biotin |
| Formulation | PBS with no preservative |
| Gene | ELAVL3 |
| Gene Accession No. | P26378, Q14576 |
| Gene Alias | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 3; ELAV like neuron-specific RNA binding protein 4; ELAV like RNA binding protein 4; elavC; ELAVL3; elavl3 protein; elavl4; ELAV-like protein 3; ELAV-like protein 4; elav-related C; elav-related D; elrc; elrd; embryonic lethal, abnormal vision-related C; embryonic lethal, abnormal vision-related D; etID309927.3; Hu antigen C; Hu antigen D; hu-antigen C; Hu-antigen D; HuC; HUCL; hud; id:ibd1248; Paraneoplastic cerebellar degeneration-associated antigen; paraneoplastic encephalomyelitis antigen HuD; Paraneoplastic limbic encephalitis antigen 21; PLE21; PNEM; protein elrD; ribonucleoprotein; RNA-binding protein HuD; wu:fb77b03; wu:fc24h01; zHuC |
| Gene Symbols | ELAVL3, ELAVL4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.