Learn More
Cellular Engineering Technologies Cryopreserved vial of human adipose-derived mesenchymal stem cells (500,000 cells)
Supplier:  Cellular Engineering Technologies CR1004500
Encompass_Preferred
Cells derived from adult human lipoaspirate tissue collected during elective surgical liposuction procedures and then programmed using our patented virus-free method using episomal DNA and our proprietary mix of vectors, excluding Myc and Lin28 transcription factors. Adipose-derived mesenchymal cells (AdMSCs) are the most commonly used mesenchymal stem cells for clinical applications.The cell line was validated for pluripotency based on colony morphology, alkaline phosphatase expression, and expression of SSEA-4. Cells are free of Mycoplasma and exhibit classical morphology and growth characteristics. AdMSCs have shown significant promise in treating autoimmune and neurodegenerative diseases, vascular and metabolic diseases, bone and cartilage regeneration, and wound defects. These cells have been reported to differentiate into many different lineages, including chondrogenic, osteogenic, adipogenic, and neural cells
Catalog No. NC2753008