Learn More
KACTUS BIOSYSTEMS HUM CD43 PROTN CD4-HM143 100UG
Supplier:  KACTUS BIOSYSTEMS CD4HM143100UG
Encompass_Preferred
Human CD43 Protein (CD4-HM143)Express System: HEK293Product Tag: C-HisPurity: > 95% as determined by Tris-Bis PAGE;> 95% as determined by HPLCExact Sequence: Ser20-Arg253
Catalog No. NC2372851
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.